missing translation for 'onlineSavingsMsg'
Learn More

Flavoxate hydrochloride, MedChemExpress

Product Code. 16448366
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 16448366

Brand: Medchem Express HYB0549A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flavoxate hydrochloride is a potent and competitive phosphodiesterase (PDE) inhibitor. Flavoxate hydrochloride is an antispasmodic agent and muscarinic mAChR antagonist. Flavoxate hydrochloride shows moderate calcium antagonistic activity and local anesthetic effect. Flavoxate hydrochloride can be used for the research of overactive bladder (OAB) and lower urinary tract infections.

Flavoxate hydrochloride is a potent and competitive phosphodiesterase (PDE) inhibitor. Flavoxate hydrochloride is an antispasmodic agent and muscarinic mAChR antagonist. Flavoxate hydrochloride shows moderate calcium antagonistic activity and local anesthetic effect. Flavoxate hydrochloride can be used for the research of overactive bladder (OAB) and lower urinary tract infections.

Specifications

Chemical Name or Material Flavoxate hydrochloride
CAS 3717-88-2
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClNO4
Quantity 100 mg
Synonym Rec-7-0040 DW61
Solubility Information H2O : 5 mg/mL (11.68 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (7.78 mM; Need ultrasonic)
SMILES O=C(C1=C2C(C(C(C)=C(C3=CC=CC=C3)O2)=O)=CC=C1)OCCN4CCCCC4.Cl
Molecular Weight (g/mol) 427.92
Formula Weight 427.92
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.