missing translation for 'onlineSavingsMsg'
Learn More

Flavopiridol, MedChemExpress

Product Code. 16432768
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432768

Brand: Medchem Express HY10005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively.

Specifications

Chemical Name or Material Flavopiridol
CAS 146426-40-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20ClNO5
Quantity 10 mg
Synonym HMR-1275 Alvocidib L86-8275
Solubility Information DMSO : 33.33 mg/mL (82.94 mM; Need ultrasonic)
SMILES O=C1C2=C(C=C(C([C@]3([H])[C@H](O)CN(C)CC3)=C2OC(C4=CC=CC=C4Cl)=C1)O)O
Molecular Weight (g/mol) 401.84
Formula Weight 401.84
Percent Purity 95.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.