missing translation for 'onlineSavingsMsg'
Learn More

Flavone, 99%

Product Code. 11480194
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480194 25 g 25g
11460194 1 g 1g
11470194 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480194 Supplier Thermo Scientific Alfa Aesar Supplier No. A13627.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 525-82-6 | C15H10O2 | 222.243 g/mol

Flavone is a potentially useful biochemical for cytochrome P450 studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavone is a potentially useful biochemical for cytochrome P450 studies.

Solubility
Soluble in acetone (25 mg/ml), methanol, alcohol, and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers.Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-82-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00006825
InChI Key VHBFFQKBGNRLFZ-UHFFFAOYSA-N
Synonym flavone, 2-phenylchromone, 2-phenyl-4h-chromen-4-one, 2-phenyl-4-chromone, 2-phenyl-4-benzopyron, 2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2-phenyl, asmacoril, chromocor, cromaril
PubChem CID 10680
ChEBI CHEBI:42491
IUPAC Name 2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2

Specifications

Melting Point 95°C to 98°C
Odor Odorless
Quantity 25 g
Beilstein 157598
Merck Index 14,4092
Solubility Information Soluble in acetone (25mg/ml),methanol,alcohol,and chloroform. Insoluble in water.
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Flavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.