missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flavivirus Envelope Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119552
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
The Flaviviridae are a family of viruses that are primarily spread through arthropod vectors (mainly ticks and mosquitoes). Flaviviridae have monopartite, linear, single-stranded RNA genomes of positive polarity, 9.6- to 12.3-kilobase in length. Virus particles are enveloped and spherical, about 40-60 nm in diameter.
Specifications
| Flavivirus Envelope | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| 1.05 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Flavivirus Envelope protein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction