missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Flavivirus Capsid Protein Polyclonal antibody specifically detects Flavivirus Capsid Protein in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Flavivirus Capsid Protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | C protein, Capsid protein C, Core protein, Flavivirus group Capsid |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Flavivirus Capsid Protein. The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?