missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flavin reductase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583580
This item is not returnable.
View return policy
Description
Immunogen sequence: DVDKTVAGQD AVIVLLGTRN DLSPTTVMSE GARNIVAAMK AHGVDKVVAC TSAFLLWDPT KVPPRLQAVT DDH Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 93%.
The final step in heme metabolism in mammals is catalyzed by the cytosolic biliverdin reductase enzymes A and B (EC 1.3.1.24).
Specifications
| Flavin reductase | |
| Polyclonal | |
| Unconjugated | |
| BLVRB | |
| Biliverdin reductase B; biliverdin reductase B (flavin reductase (NADPH)); biliverdin-IX beta-reductase; BLVRB; BVRB; BVR-B; epididymis secretory protein Li 10; flavin reductase (NADPH); FLR; FR; GHBP; green heme-binding protein; HEL-S-10; NADPH; NADPH-dependent diaphorase; NADPH-flavin reductase; SDR43U1; short chain dehydrogenase/reductase family 43U, member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P30043 | |
| BLVRB | |
| Recombinant protein corresponding to Human BLVRB. Recombinant protein control fragment (Product #RP-98899). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction