missing translation for 'onlineSavingsMsg'
Learn More

Flavin Adenine Dinucleotide Disodium Salt Hydrate, ≥93% (dry basis)

Product Code. 15825288 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Plastic bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15825288 500 mg Plastic bottle 500mg
15875138 1 g Plastic bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825288 Supplier Thermo Scientific Supplier No. J15785.ME

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1891059-93-0 | C27H31N9Na2O15P2 | 829.52 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1891059-93-0
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5 '-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
PubChem CID 131675332
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Physical Form Crystalline Powder
Quantity 500 mg
IUPAC Name disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(2R,3S,4S)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl phosphonato]oxy}phosphonate
Formula Weight 829.51 (anhydrous)
Percent Purity ≥93%
Packaging Plastic bottle
Assay Dry basis
Chemical Name or Material Flavin Adenine Dinucleotide Disodium Salt Hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.