Learn More
Grant Instruments™ Flat Platform With Non-Slip Mat for PSU20i
Description
- For use with PSU 10i Orbital Shaking Platform
- Accommodates low profile vessesl such as Petri dishes
- Black rubber pads provide nonslip surface for vessels on the platform.
Specifications
Specifications
| Material | Rubber |
| Dimensions (L x W) | 480 x 380 mm |
| Description | Flat platform |
| For Use With (Equipment) | PSU-20i |
| No. of Shelves | Flat platform |