Learn More
Lenz Laborglasinstrumente™ Flat Flange Lids With Center Neck
Brand: Lenz Laborglasinstrumente™ 06100201
Code : 50
Additional Details : Weight : 1.00000kg
Specifications
Borosilicate Glass, Duran Glass | |
80 mm | |
Flange lid |
Reaction vessels | |
Laboratory Flange 100 | |
Center Neck (NS): 14/23 |