Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Two Angled Side Necks, and One Vertical Side Neck
Brand: Lenz Laborglasinstrumente™ 06104544
Code : 50
Additional Details : Weight : 1.00000kg
Specifications
Borosilicate Glass, Duran Glass | |
Laboratory Flange 200 | |
Center Neck (NS): 29/32 |
Reaction vessels | |
Flange lid |