Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Two Angled Side Necks, and One Vertical Side Neck
Brand: Lenz Laborglasinstrumente™ 06104264
Code : 50
Additional Details : Weight : 0.99000kg
Specifications
Borosilicate Glass, Duran Glass | |
Laboratory Flange 100 | |
Center Neck (NS): 45/40 |
Reaction vessels | |
Flange lid |