missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimble Chase™ Flat Bottom HPLC Reservoir with Graduations
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
Description
- Recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents
- Plastic safety-coating blocks UV light up to 385nm, preventing photo-degradation of light-sensitive reagents
- Safety coating helps to retain glass fragments and allows a reasonable amount of time for the safe disposal of liquid contents
- Used with internal pressures up to 0.4 bar (6 psig) for helium sparging and blanketing of the mobile phase
- Operating Pressure: -1.0 to 0.4 bar (-14.5 psig to 6 psig)
- Chemically inert glass prevents leaching of any extractables into the mobile phase solvents
- Supplied with GL-45 screw thread
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| For Use With (Application) | HPLC |
| Material | Borosilicate Glass (Class A, Type I) |
| No. per Case | 1Each |
| Height (Metric) | 275 mm |
| Product Type | Flat Bottom with Graduations |
| Diameter (Metric) | 130 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction