missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Flat Bottom Flasks, Wide Neck

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07050073

 View more versions of this product

Product Code. 11953831

  • £558.00 / Pack of 10
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN ISO 24450
  • With beaded rim
  • Material: DURAN™
Specifications

Specifications

2 L
Duran Glass
230 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers