Learn More
JULABO™ Flat Bath Cover for PURA™ Water Baths
Click to view available options
For Use With (Equipment):
PURA™ 10
PURA™ 14
PURA™ 22
PURA™ 30
Type:
1 opening, 190 mm dia.
10 openings, 115 mm dia.
2 openings, 190 mm dia.
3 openings, 190 mm dia.
4 openings, 92 mm dia.
6 openings, 115 mm dia.
6 openings, 92 mm dia.
8 openings, 92 mm dia.
Unit Size:
Each
Description
- Accessory for PURA™ water bath
- With appropriate sets of rings
Specifications
Specifications
| Description | Flat bath cover |
| For Use With (Equipment) | PURA™ 30 |
| Type | 3 openings, 190 mm dia. |