missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Silica

Product Code. 16350146 Shop All Buchi Products
Click to view available options
Includes:
10 Pc.
12 Pc.
18 Pc.
20 Pc.
5 Pc.
6 Pc.
Bed Weight (Metric):
4 g
12 g
25 g
40 g
80 g
220 g
330 g
Unit Size:
Pack of 10
Pack of 12
Pack of 18
Pack of 20
Pack of 5
Pack of 6
This item is not returnable. View return policy

Product Code. 16350146

Brand: BUCHI 140000024

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters

Specifications

Material Polypropylene
Type Silica
Includes 12 Pc.
Resolution 40 to 63 μm
Bed Weight (Metric) 40 g
Column Type Silica
For Use With (Application) Basic Applications, Polar Compounds
For Use With (Equipment) Any Flash System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.