missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Diol

Product Code. 16370186 Shop All Buchi Products
Change view
Click to view available options
Size (Metric) Tube Rack:
120 g
Includes:
2 psc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Tube Rack Includes
16370186 120 g 2 psc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16370186 Supplier BUCHI Supplier No. 140000072
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Type Diol
Includes 2 psc
Resolution 40 to 60 μm
Size (Metric) Tube Rack 120 g
Column Type Diol
For Use With (Application) Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
For Use With (Equipment) Any Flash System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.