Learn More
BUCHI FlashPure Chromatography Solid Loader
Product Code. 16491374 Shop All Buchi Products
Change view
Click to view available options
Size (Metric) Tube Rack:
15 g
Unit Size:
1 set
Product Code. Size (Metric) Tube Rack unitSize
16491374 15 g 1 set
1 options
Product Code. 16491374 Supplier BUCHI Supplier No. 11068975
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
TRUSTED_SUSTAINABILITY
Specifications
Type Chromatography Loader
Includes Adapter Set, Sleeve, 20 Tubes, and 40 Frits
Size (Metric) Tube Rack 15 g
For Use With (Equipment) Pure Chromatography Systems