missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System DNA Cassettes
Description
- Get results in 5 minutes or less
- Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Concentration | 2.2% |
| Separation Range | 10 to 1000 bp |
| Wells | 12 + 1-well |
| Formulation | 2.2% Agarose |
| For Use With (Application) | Electrophoresis |
| Kit Contents | Agarose Gel, Stain and Buffer |
| Material | Agarose |
| Quantity | 9 Pack |
| Storage Requirements | 18° to 26°C |
| Length (English) Cassette | 4.5 in. |
| Show More |
Safety and Handling
Product Identifier
- FlashGel System
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contain: Tetrabutylammonium hydroxide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?