missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ FlashGel™ System DNA Cassettes
Cassettes optimized for DNA migration. Lonza™ FlashGel™ System DNA Cassettes separate DNA quickly and DNA migration can be seen as it happens. Separates DNA in 2 to 7 minutes.
Brand: Lonza™ 57029
Code : NEW
Additional Details : Weight : 0.90000kg
Description
- Get results in 5 minutes or less
- Contain pre-stained agarose gels and buffer, minimizing user exposure to potentially mutagenic stains
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
1.2% | |
50 to 4000 bp | |
1.2% Agarose | |
Agarose Gel, Stain and Buffer | |
9 Pack | |
4.5 in. | |
11.5 cm | |
4.2 in. | |
8.4 cm | |
1.7 cm |
DNA Cassettes | |
16 + 1-well | |
Electrophoresis | |
Agarose | |
18° to 26°C | |
2.7 in. | |
7 cm | |
10.7 cm | |
0.6 in. |