missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flash Point Standard - Pensky Martens, ASTM D 93 Proc. A (nominal value: 184.0 °C), Paragon Scientific™
Click to view available options
Quantity:
3 x 80 mL
Unit Size:
Pack of 3
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Packaged in 3 X 80mL bottles
- One bottle per test ensures zero contamination / high end volatile loss
- Fully traceable to International Standards
- Supplied with MSDS
- Tamper evident security packaging
- 12 Month shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 3 x 80 mL |
Nominal values quoted. Certified Values will vary from batch to batch.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction