missing translation for 'onlineSavingsMsg'
Learn More

FlAsH-EDT2, TRC

Product Code. 30407729
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407729 50 mg
30391386 25 mg
30402548 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407729 Supplier Toronto Research Chemicals Supplier No. TRCF33520050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212118-77-9
Molecular Formula C24 H18 As2 O5 S4
Molecular Weight (g/mol) 664.5
IUPAC Name 2-[4,5-bis(1,3,2-dithiarsolan-2-yl)-3-hydroxy-6-oxoxanthen-9-yl]benzoic acid
SMILES OC(=O)c1ccccc1C2=C3C=CC(=O)C(=C3Oc4c2ccc(O)c4[As]5SCCS5)[As]6SCCS6

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H18As2O5S4/c27-17-7-5-15-19(13-3-1-2-4-14(13)24(29)30)16-6-8-18(28)21(26-34-11-12-35-26)23(16)31-22(15)20(17)25-32-9-10-33-25/h1-8,27H,9-12H2,(H,29,30)
Formula Weight 663.8469
Percent Purity >95%
Chemical Name or Material FlAsH-EDT2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.