Description
- Includes: Flange Nut, and Pressure Spring
Specifications
Specifications
| Connection Type | Flange-screwed |
| Description | Compatible with Glass Assembly V/C/A |
| Material | PBT |
| Type | Flange Nut |
| For Use With (Application) | Securing V/C/A/HP Condenser |
| For Use With (Equipment) | Rotavap R-300, R-210, R-215 |