Learn More
Key Industrial Equipment™ Flammable material cabinet, yellow, 900mm x 915mm x 459mm
Brand:  Key Industrial Equipment™ 1956R035N
Encompass_Preferred
Product Code. 12836919
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
A double door cabinet robustly constructed from 0. 9mm thick steel. Designed to maintain its integrity during the early minutes of exposure to fire. To ensure access to hazardous chemicals is limited to authorised personnel, the doors are opened by a chrome handle and secured with a 3-point locking system, supplied with 2 keys for convenience. Has perforated height adjustable shelf trays to allow spillages to drain into a liquid tight sump tray. 80mm deep removable sump tray will contain any spillage. The cabinet is also fully sealed at bottom with point welding all around, to further prevent leakage at ground level. Finish: high visibility epoxy powder coated yellow and labelled to UK and European standards, with an appropriate 'Highly Flammable' warning label. Comply with HSG51, COSHH, DSEAR.
  • One shelf
  • Dimensions (H x W x D) : 900mm x 915mm x 459mm