missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flamingo homolog 1/CELSR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Flamingo homolog 1/CELSR3 |
|---|---|
| Dilution | Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686695
|
Novus Biologicals
NBP2-38975-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18105509
|
Novus Biologicals
NBP2-38975 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flamingo homolog 1/CELSR3 Polyclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Flamingo homolog 1/CELSR3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9NYQ7 | |
| 1951 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LDSLSRSSNSREQLDQVPSRHPSREALGPLPQLLRAREDSVSGPSHGPSTEQLDILSSILASFNSSALSSVQSSSTP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11RESDA1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2 | |
| CELSR3 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title