missing translation for 'onlineSavingsMsg'
Learn More

Flamingo homolog 1/CELSR3 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30733857 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733857 Supplier Novus Biologicals Supplier No. AF7278M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Flamingo homolog 1/CELSR3 Polyclonal antibody specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flamingo homolog 1/CELSR3
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11RESDA1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2
Host Species Goat
Immunogen E. coli-derived recombinant human CELSR3, Tyr531-Ser711, Accession # Q9NYQ7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1951
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.