missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flamingo homolog 1/CELSR3 Antibody (763103), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7278APCCY7
This item is not returnable.
View return policy
Description
Flamingo homolog 1/CELSR3 Monoclonal antibody specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry
Specifications
| Flamingo homolog 1/CELSR3 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 763103 | |
| Flow Cytometry | |
| Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11RESDA1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2 | |
| E. coli-derived recombinant human CELSR3, Val531-Ser711, Accession # Q9NYQ7 | |
| 0.1 mL | |
| GPCR | |
| 1951 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction