Learn More
Thermo Scientific™ Flame Filter for Solvents
To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21 °C
Brand: Thermo Scientific™ 50042626
2072.17 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Code : 51
Additional Details : Weight : 0.10000kg
Description
To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21 °C
- To be connected to vacuum port at rear wall, is recommended for applications with solvents with ignition temperatures of < 21 °C.
Specifications
Flame Filter | |
Vacutherm VT 6000 M/P-BL Oven, Vacuum Tubing |