missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Flagellin Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Binds to human & mouse TLR5. Activation of TLR5 in human epithelial cell assays based on NF-kB luciferase fusions, on expression of IL-8 (CXCL8), CCL20, inducible nitric oxide synthase, beta-defensin 2 & matrilysin (MMP-7). Maturation of dendritic cells in in vitro & in vivoassays. Adjuvant activity on foreign antigens delivered by subcutaneous & intranasal routes in mice. TLR5 can be stimulated in vitro by Flagellin 5-50ng/mL & in vivo at a starting concentration of 0.1-1 ug/mouse.)
Specifications
Specifications
| Accession Number | P06179 |
| Concentration | LYOPH |
| For Use With (Application) | Bioactivity, ELISA |
| Formulation | Lyophilized from PBS |
| Quantity | 100 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.1 EU/μg purified protein (LAL test; Lonza). |
| Reconstitution | Reconstitute with 100 ul sterile water (1mg/ml after reconstitution) |
| Research Category | Immunology, Innate Immunity |
| Purity or Quality Grade | >90% SDS-PAGE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction