missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Flagellin Native Protein
Description
Binds to human & mouse TLR5. Activation of TLR5 in human epithelial cell assays based on NF-kB luciferase fusions, on expression of IL-8 (CXCL8), CCL20, inducible nitric oxide synthase, beta-defensin 2 & matrilysin (MMP-7). Maturation of dendritic cells in in vitro & in vivo assays. Adjuvant activity on foreign antigens delivered by subcutaneous & intranasal routes in mice. TLR5 can be stimulated in vitro by Flagellin (high Purity) at 5-50ng/mL & in vivo at a starting concentration of 0.1-1ug/mouse.
Specifications
Specifications
| Accession Number | P06179 |
| Concentration | LYOPH |
| For Use With (Application) | Bioactivity, ELISA |
| Formulation | Lyophilized from PBS |
| Quantity | 10 μg |
| Storage Requirements | Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.01 EU/μg purified protein (LAL test; Lonza). |
| Reconstitution | Reconstitute with 100 ul sterile water (0.1mg/ml after reconstitution) |
| Research Category | Immunology, Innate Immunity |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?