missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Flagellin Monoclonal Antibody (06)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528968
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Listeria monocytogenes Listeria monocytogenes flagellin/FlaA.
Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella.
Specifications
| Flagellin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 06 | |
| Unconjugated | |
| flaF; Flagellin; fliC; hag; hase 1-A flagellin; SPA0911 | |
| Recombinant Listeria monocytogenes Listeria monocytogenes flagellin/FlaA protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction