missing translation for 'onlineSavingsMsg'
Learn More

Flagellin Antibody (121), Biotin, Novus Biologicals™

Product Code. 18366470 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366470 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366470 Supplier Novus Biologicals Supplier No. NBP290335B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Flagellin Monoclonal specifically detects Flagellin in Bacteria samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flagellin
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 121
Conjugate Biotin
Dilution ELISA
Formulation PBS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Listeria monocytogenes Listeria monocytogenes flagellin/FlaA
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.