missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Flagellin Monoclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Flagellin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 06 |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Listeria monocytogenes Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287). |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?