Learn More
Invitrogen™ FLAD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589731
Description
Immunogen sequence: ETSLAQYSLT QLCVGFNGGK DCTALLHLFH AAVQRKLPDV PNPLQILYIR SISPFPELEQ FLQDTIKRYN LQMLEAEGSM KQALGELQAR HPQLEAVLMG TRRTDPYSCS LCPFSPTDPG WPAFMRINPL LDWTYRDIWD FLRQLFVPYC ILYDRGYTSL GSRENTVRNP ALKCLSPGGH PTYRPAYLLE NEEEERNSRT; Positive Samples: U-87MG, Mouse brain; Cellular Location: Cytoplasm, Mitochondrion matrix.
FLAD1 encodes the enzyme that catalyzes adenylation of flavin mononucleotide (FMN) to form flavin adenine dinucleotide (FAD) coenzyme.
Specifications
| FLAD1 | |
| Polyclonal | |
| Unconjugated | |
| FLAD1 | |
| A930017E24Rik; fa92c06; FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD synthetase; FAD1; FAD1 flavin adenine dinucleotide synthetase homolog; Fad1, flavin adenine dinucleotide synthetase, homolog; FADS; FAD-synthetase; FLAD1; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; flavin adenine dinucleotide synthetase 1; FMN adenylyltransferase; hypothetical protein LOC506682; LSMFLAD; Molybdenum cofactor biosynthesis protein-like region; PP591; RFad1, flavin adenine dinucleotide synthetase, homolog (yeast); RP11-307C12.7; wu:fa92c06; zgc:91843 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 319945, 80308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8NFF5, Q8R123 | |
| FLAD1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 388-587 of human FLAD1 (NP_0794833). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.