Learn More
Liebherr Refrigerator FKv3643 upright, 5 grid shelves plastics coated mechanic control system 220-240V 50Hz 335L capacity +1°C to +15°C dynamic cooling system, free standing, glazed door Liebherr
Brand:  Liebherr FKV 3643
Refrigerator FKv3643 upright, 5 grid shelves plastics coated mechanic control system 220-240V 50Hz 335L capacity +1°C to +15°C dynamic cooling system, free standing, glazed door Liebherr
Code : 03
Additional Details : Weight : 74.00000kg
Product Code. 15103243