missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBPL Mouse anti-Human, Mouse, Clone: 5D9A9, Proteintech
Mouse Monoclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | FKBPL |
|---|---|
| Clone | 5D9A9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.Specifications
| FKBPL | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DIR1, FK506 binding protein like, FKBPL, NG7, WISP39 | |
| Fkbpl | |
| IgG1 | |
| Caprylic Acid Precipitation | |
| Antibody |
| 5D9A9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| O35450, Q9UIM3 | |
| 56299, 63943 | |
| FKBPL Fusion Protein Ag0112 | |
| Primary | |
| -20°C | |
| FKBPL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title