missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £484.00
Specifications
| Antigen | FKBP9 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290633
|
Novus Biologicals
NBP2-58218 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621498
|
Novus Biologicals
NBP2-58218-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP9 Polyclonal specifically detects FKBP9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| FKBP9 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 11328 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FDAELIVKNMFTNQDRNGDGKVTAEEFKL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| 63 kDa FK506-binding protein, DKFZp586B1723, FK506 binding protein 9, 63 kDa, FKBP60, FKBP63, FKBP-9, MGC126772,63 kDa FKBP, MGC138258, peptidyl-prolyl cis-trans isomerase FKBP9, PPIase, rotamase | |
| FKBP9 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title