missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP8 Recombinant Rabbit Monoclonal Antibody (23GB5890), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552940
This item is not returnable.
View return policy
Description
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
Specifications
| FKBP8 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FKBP8 | |
| 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23770 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5890 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14318 | |
| FKBP8 | |
| A synthesized peptide derived from human FKBP38 (1-46AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction