missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11173-1-AP-150UL
This item is not returnable.
View return policy
Description
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.Specifications
| FKBP8 | |
| Polyclonal | |
| Unconjugated | |
| FKBP8 | |
| 38 kDa FK506 binding protein, 38 kDa FKBP, FK506 binding protein 8, FK506 binding protein 8, 38kDa, FKBP 38, FKBP 8, FKBP38, FKBP8, FKBPr38, hFKBP38, PPIase FKBP8, Rotamase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14232, 23770 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35465, Q14318 | |
| Fkbp8 | |
| FKBP8 Fusion Protein Ag1649 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction