missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580637
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FKBP7/Rotamase.
The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.
Specifications
| FKBP7 | |
| Polyclonal | |
| Unconjugated | |
| FKBP7 | |
| 23 kDa FK506-binding protein; 23 kDa FKBP; 23kDa; FK506 binding protein 7; FK506-binding protein 23; FK506-binding protein 7; FKBP23; FKBP-23; Fkbp7; FKBP-7; Peptidyl-prolyl cis-trans isomerase FKBP7; PPIase; PPIase FKBP7; rotamase; UNQ670/PRO1304 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y680 | |
| FKBP7 | |
| Recombinant Human FKBP7/Rotamase protein (Met1-Gln218). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction