missing translation for 'onlineSavingsMsg'
Learn More

FKBP7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

£305.00 - £435.00

Specifications

Antigen FKBP7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18428220
View Documents
Novus Biologicals
NBP1-83889-25ul
25 μL
£305.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18286198
View Documents
Novus Biologicals
NBP1-83889
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FKBP7 Polyclonal specifically detects FKBP7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

FKBP7
Polyclonal
Rabbit
Human, Mouse
EC 5.2.1.8, FK506 binding protein 7, FK506-binding protein 7FK506-binding protein 23,23 kDa FK506-binding protein, FKBP-23, FKBP23PPIase, FKBP-7, MGC9420, peptidyl-prolyl cis-trans isomerase FKBP7,23 kDa FKBP, PPIase FKBP7, rotamase
FKBP7
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
51661
This antibody was developed against Recombinant Protein corresponding to amino acids:AEGKIPPDATLIFEIELYAVTKGPRSIETFKQIDMDNDRQLSKAEINLYLQREFEKDEKPRDKSYQDAVLEDIFKKNDHDGDGFISPKEYNVYQHDEL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.