missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FKBP7 Monoclonal antibody specifically detects FKBP7 in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Clone | 005 |
| Gene Alias | EC 5.2.1.8, FK506 binding protein 7, FK506-binding protein 7FK506-binding protein 23,23 kDa FK506-binding protein, FKBP-23, FKBP23PPIase, FKBP-7, MGC9420, peptidyl-prolyl cis-trans isomerase FKBP7,23 kDa FKBP, PPIase FKBP7, rotamase |
| Gene ID (Entrez) | 51661 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?