missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FKBP6 Recombinant Rabbit Monoclonal Antibody (K01_2M43)
GREENER_CHOICE

Product Code. 30828621
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828621 100 μL
30829590 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828621 Supplier Invitrogen™ Supplier No. MA564065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

FKBP6 Recombinant Monoclonal Antibody for IHC (P), Western Blot

FKBP6 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP6 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin.This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2M43
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Fkbp6
Gene Accession No. D3ZQF4, O75344, Q91XW8
Gene Alias 1700008G22Rik; 36 kDa FK506 binding protein; 36 kDa FK506-binding protein; 36 kDa FKBP; 36kDa; As; AU017274; D5Ertd724e; FK506 binding protein 6; FK506 binding protein 6, 36kDa; FK506-binding protein 6; FK506-binding protein 6 (36kD); FKBP prolyl isomerase 6; FKBP prolyl isomerase family member 6 (inactive); Fkbp36; FKBP-36; FKBP6; FKBP-6; Immunophilin FKBP36; Inactive peptidyl-prolyl cis-trans isomerase FKBP6; Inactive PPIase FKBP6; MGC87179; peptidyl-prolyl cis-trans isomerase FKBP6; PPiase; PPIase FKBP6; rotamase
Gene Symbols Fkbp6
Host Species Rabbit
Immunogen A synthetic peptide of human FKBP6.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288597, 8468, 94244
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.