missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FKBP6 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
FKBP6 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP6 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin.This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.
Specifications
Specifications
| Antigen | FKBP6 |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Fkbp6 |
| Gene Accession No. | D3ZQF4, O75344, Q91XW8 |
| Gene Alias | 1700008G22Rik; 36 kDa FK506 binding protein; 36 kDa FK506-binding protein; 36 kDa FKBP; 36kDa; As; AU017274; D5Ertd724e; FK506 binding protein 6; FK506 binding protein 6, 36kDa; FK506-binding protein 6; FK506-binding protein 6 (36kD); FKBP prolyl isomerase 6; FKBP prolyl isomerase family member 6 (inactive); Fkbp36; FKBP-36; FKBP6; FKBP-6; Immunophilin FKBP36; Inactive peptidyl-prolyl cis-trans isomerase FKBP6; Inactive PPIase FKBP6; MGC87179; peptidyl-prolyl cis-trans isomerase FKBP6; PPiase; PPIase FKBP6; rotamase |
| Gene Symbols | Fkbp6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?