missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP6 Polyclonal Antibody

FKBP6 Polyclonal Antibody
Brand: Thermo Scientific PA563926
This item is not returnable.
View return policy
Description
FKBP6 Polyclonal Antibody for Western Blot, IHC
Specifications
| FKBP6 | |
| Polyclonal | |
| Unconjugated | |
| Fkbp6 | |
| 1700008G22Rik; 36 kDa FK506 binding protein; 36 kDa FK506-binding protein; 36 kDa FKBP; 36kDa; As; AU017274; D5Ertd724e; FK506 binding protein 6; FK506 binding protein 6, 36kDa; FK506-binding protein 6; FK506-binding protein 6 (36kD); FKBP prolyl isomerase 6; FKBP prolyl isomerase family member 6 (inactive); Fkbp36; FKBP-36; FKBP6; FKBP-6; Immunophilin FKBP36; Inactive peptidyl-prolyl cis-trans isomerase FKBP6; Inactive PPIase FKBP6; MGC87179; peptidyl-prolyl cis-trans isomerase FKBP6; PPiase; PPIase FKBP6; rotamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75344 | |
| Fkbp6 | |
| Recombinant protein corresponding to Human FKBP6. Recombinant protein control fragment (Product #RP-103389). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction