missing translation for 'onlineSavingsMsg'
Learn More

FKBP6 Antibody (S05-4G2), Novus Biologicals™

Product Code. 18626973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18626973 50 μg 50µL
18697823 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18626973 Supplier Novus Biologicals Supplier No. NBP31986550ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FKBP6 Monoclonal antibody specifically detects FKBP6 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP6
Applications Western Blot
Classification Monoclonal
Clone S05-4G2
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase
Host Species Rabbit
Immunogen A synthetic peptide of human FKBP6 (Uniprot # O75344)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 8468
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.