missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FKBP6 Monoclonal antibody specifically detects FKBP6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | FKBP6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S05-4G2 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human FKBP6 (Uniprot # O75344) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?