missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP6 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97729PE
This item is not returnable.
View return policy
Description
FKBP6 Polyclonal antibody specifically detects FKBP6 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| FKBP6 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase | |
| Produced in rabbits immunized with E. coli-derived Human FKBP6 fragment. | |
| 0.1 mL | |
| Immunology | |
| 8468 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction