missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£361.00
Specifications
| Antigen | FKBP6 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
FKBP6 Polyclonal specifically detects FKBP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FKBP6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase | |
| FKBP6 | |
| IgG | |
| Protein A purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| O75344 | |
| 8468 | |
| Synthetic peptides corresponding to FKBP6(FK506 binding protein 6, 36kDa) The peptide sequence was selected from the middle region of FKBP6. Peptide sequence FCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLL. | |
| Primary | |
| 53 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title