missing translation for 'onlineSavingsMsg'
Learn More

FKBP51/FKBP5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18375402 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375402 100 μg 100µL
18333534 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18375402 Supplier Novus Biologicals Supplier No. NBP318673100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FKBP51/FKBP5 Polyclonal antibody specifically detects FKBP51/FKBP5 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP51/FKBP5
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:500, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation Whole antiserum
Gene Accession No. Q13451
Gene Alias 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, rotamase, T-cell FK506-binding protein
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse FKBP51/FKBP5 (FK506 Binding Protein 51), using two KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein (1).
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2289
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.