Learn More
Invitrogen™ FKBP5 Monoclonal Antibody (K52044_5B2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555755
Description
Sequence of this protein is as follows: DLFEDGGIIR RTKRKGEGYS NPNEGATVEI HLEGRCGGRM FDCRDVAFTV GEGEDHDIPI GIDKALEKMQ REEQCILYLG PRYGFGEAGK PKFGIEPNAE LIYEVTLKSF EKAKESWEMD TKEKLEQAAI VKEKGTVYFK GGKYMQAVIQ YGKIVSWLEM EYGLSEKESK ASESFLLAAF LNLAMCYLKL REYTKAVECC DKALGLDSAN EKGLYRRGEA QLLMNEFESA KGDFEKVLEV NPQNKAARLQ ISMCQKKAKE HNERDRRIYA NMFKKFAEQD AKEEANKAMG KKTSEGVTNE KGTDSQAMEE
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.
Specifications
| FKBP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13451 | |
| FKBP5 | |
| Recombinant human FKBP5 (immunogen range is 141-450). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K52044_5B2 | |
| Unconjugated | |
| FKBP5 | |
| 51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 2289 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.